Skip to Content
Merck
All Photos(1)

Key Documents

347299

Sigma-Aldrich

2,6-Difluorotoluene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3F2
CAS Number:
Molecular Weight:
128.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
99%

Assay

99%

form

liquid

refractive index

n20/D 1.453 (lit.)

bp

112 °C/740 mmHg (lit.)

density

1.129 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Cc1c(F)cccc1F

InChI

1S/C7H6F2/c1-5-6(8)3-2-4-7(5)9/h2-4H,1H3

InChI key

MZLSNIREOQCDED-UHFFFAOYSA-N

General description

2,6-Difluorotoluene reacts with chlorine to give 2,6-difluorobenzyl chloride, which gets converted to 2,6-difluorobenzaldehyde through Sommelet′s reaction. Six-fold potential for internal methyl rotation in first singlet excited state and cation ground state of 2,6-difluorotoluene has been determined.

Application

2,6-Difluorotoluene was used to generate jet-cooled 2,6-difluorobenzyl radical and to investigate its vibronically resolved emission spectra.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Observation of vibronic emission spectrum of the jet-cooled 2, 6-difluorobenzyl radical.
Lee SK and Baek DY.
The Journal of Physical Chemistry A, 104(22), 5219-5221 (2000)
Methyl Group Internal Rotation in 2, 6-Difluorotoluene (S1) and 2, 6-Difluorotoluene+ (D0).
Walker RA, et al.
The Journal of Physical Chemistry, 99(33), 12422-12433 (1995)
Preparation of 2, 6-difluoro-n-alkylbenzenes from 1, 3-difluorobenzene Transformation of 2, 6-difluorotoluene to the corresponding benzaldehyde via benzyl chloride.
Malykhin EV and Shteingarts VD.
Journal of Fluorine Chemistry, 9(1), 19-20 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service