Polyvinylpyrrolidone
Synonym(s):
PVP, Polyvidone, Povidone, PVPP, Polyvinylpyrrolidone, cross-linked, Polyvinylpyrrolidone, Polyvinylpolypyrrolidone
Linear Formula:
(C6H9NO)n
CAS Number:
Products
Shipping
Product Category
Brand
Feature
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Polymer Composition
Polymer Type
API Family
Special Grade
Technique
Showing 1-20 of 23
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2