52214-U
Upchurch VHP (Very High Pressure) Fittings
VHP-205, for use with 1/16 inch (1.6mm) tubing, 10-32 thread, stainless steel
All Photos(1)
About This Item
Recommended Products
material
stainless steel
manufacturer/tradename
VHP-205
packaging
1 ea of
fitting
10-32 thread
compatibility
for use with 1/16 inch (1.6mm) tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)