Skip to Content
Merck
All Photos(1)

Documents

T4687

Sigma-Aldrich

Thimerosal

meets USP testing specifications

Synonym(s):

2-(Ethylmercuriomercapto)benzoic acid sodium salt, Ethylmercurithiosalicylic acid sodium salt, Mercury-([o-carboxyphenyl]thio)ethyl sodium salt, Sodium ethylmercurithiosalicylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(C2H5HgS)C6H4CO2Na
CAS Number:
Molecular Weight:
404.81
Beilstein:
8169555
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

synthetic (Organic)

Agency

USP/NF
meets USP testing specifications

form

powder

mp

234-237 °C (dec.) (lit.)

cation traces

Hg: ≤0.70% (ions)

Mode of action

enzyme | inhibits

SMILES string

[Na+].CC[Hg]Sc1ccccc1C([O-])=O

InChI

1S/C7H6O2S.C2H5.Hg.Na/c8-7(9)5-3-1-2-4-6(5)10;1-2;;/h1-4,10H,(H,8,9);1H2,2H3;;/q;;2*+1/p-2

InChI key

RTKIYNMVFMVABJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Thimerosal is a sulfhydryl (thiol) oxidizing reagent capable of inhibiting a wide range of sulfhydryl-dependent enzymes and proteins such as PI3-kinase-dependent methionine synthase, glutamate transport receptors, and Na++K+ dependent ATPases.

Other Notes

As part of the corporation global plan to support the Minamata Convention eliminating mercury-containing compounds, as of January 2021 customers will be required to provide an End-Use Declaration. Sales of thimerosal will be restricted for approved applications only.
Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pan-Chyr Yang et al.
Journal of the Formosan Medical Association = Taiwan yi zhi, 111(6), 333-339 (2012-07-04)
This study evaluated the immune response elicited by two formulations of an AS03(A)-adjuvanted H5N1 A/Indonesia/05/2005 prepandemic influenza vaccine, developed using manufacturing processes with or without thiomersal. In addition, it also evaluated compliance to the Centre for Biologics Evaluation and Research
Dorota Mrozek-Budzyn et al.
Neurotoxicology and teratology, 34(6), 592-597 (2012-10-17)
Despite the common use of Thimerosal as a preservative in childhood vaccines since the 1930s, there are not many studies on ethylmercury toxicokinetics and toxicodynamics in infants. The knowledge of ethylmercury's potential adverse effects is derived mostly from parallel methylmercury
Alicia Bazzano et al.
American journal on intellectual and developmental disabilities, 117(3), 233-242 (2012-06-22)
Although the assertion of a link between vaccines and autism has been scientifically rejected, the theory continues to be popular and may influence the attitudes of parents of children with autism spectrum disorders. The authors sought to assess how often
Alberto Eugenio Tozzi et al.
Developmental medicine and child neurology, 54(9), 843-848 (2012-05-18)
The aim of this article was to explore the effect of duration of breastfeeding on neurocognitive development. The long-term effect of breastfeeding on neurodevelopment was examined through a battery of neuropsychological tests in 1403 children (693 females, 710 males; mean
Janet K Kern et al.
International journal of environmental research and public health, 10(8), 3771-3800 (2013-08-24)
Autism spectrum disorder (ASD) is a neurological disorder in which a significant number of the children experience a developmental regression characterized by a loss of previously acquired skills and abilities. Typically reported are losses of verbal, nonverbal, and social abilities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service