Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702782

Sigma-Aldrich

Anti-FOXP1 Antibody, clone 2Q1R5, Rabbit Monoclonal

Synonym(s):

12CC4, HSPC215, MFH, QRF1, hFKH1B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

2Q1R5, monoclonal

form

liquid

mol wt

70-90 kDa

species reactivity

rat, human, rat

concentration

0.5mg/ml

technique(s)

immunofluorescence: 1:50 - 1:200
immunohistochemistry: 1:100 - 1:500
western blot: 1:1000 - 1:3700

color

colorless

isotype

IgG

immunogen sequence

SGNPSLIKNMQSSHAYCTPLNAALQASMAENSIPLYTTASMGNPTLGNLASAIREELNGAMEHTNSNESDSSPGRSPMQAVHPVHVKEEPLDPEEAEGPLS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXP1(27086)

General description

This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Forkhead box P1 protein contains both DNA-binding- and protein-protein binding-domains. This gene may act as a tumor suppressor as it is lost in several tumor types and maps to a chromosomal region (3p14.1) reported to contain a tumor suppressor gene(s). Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Jul 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 550-650 of human FOXP1 (Q9H334).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service