Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502414

Sigma-Aldrich

Anti-MAP2K7 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Dual specificity mitogen-activated protein kinase kinase 7, JNK activating kinase 2, JNK kinase 2, JNKK 2, c-Jun N-terminal kinase kinase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 47 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAP2K7(5609)

General description

Anti-MAP2K7 Antibody detects endogenous levels of total MAP2K7 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human MAP2K7.

Immunogen Range: 236-285

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Sanjana P Devi et al.
Biochimica et biophysica acta, 1840(9), 2794-2805 (2014-05-09)
Newer strategies for augmenting immune responses of pharmacologically active glucans may serve to improve the medicinal potential of these biomolecules. With this aim, the present work was focused on generating targeted high molecular size glucan particles with magnified immune response
Meiyan Sun et al.
Endocrinology, 155(8), 2787-2796 (2014-05-16)
Calcium-dependent chloride conductances have been described in chicken and human granulosa cells (GCs) and may be involved in steroidogenesis. However, the molecular identities of corresponding chloride channels remain unknown. The purpose of this study was to explore the expression and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service