Skip to Content
Merck
All Photos(1)

Key Documents

SAB1411361

Sigma-Aldrich

Anti-C2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CO2, DKFZp779M0311

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 83.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C2(717)

Related Categories

General description

Component C2 is a serum glycoprotein that functions as part of the classical pathway of the complement system. Activated C1 cleaves C2 into C2a and C2b. The serine proteinase C2a then combines with complement factor 4b to create the C3 or C5 convertase. Deficiency of C2 has been reported to associated with certain autoimmune diseases and SNPs in this gene have been associated with altered susceptibility to age-related macular degeneration. This gene localizes within the class III region of the MHC on the short arm of chromosome 6. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Additional transcript variants have been described in publications but their full-length sequence has not been determined

Immunogen

C2 (NP_000054.2, 1 a.a. ~ 752 a.a) full-length human protein.

Sequence
MGPLMVLFCLLFLYPGLADSAPSCPQNVNISGGTFTLSHGWAPGSLLTYSCPQGLYPSPASRLCKSSGQWQTPGATRSLSKAVCKPVRCPAPVSFENGIYTPRLGSYPVGGNVSFECEDGFILRGSPVRQCRPNGMWDGETAVCDNGAGHCPNPGISLGAVRTGFRFGHGDKVRYRCSSNLVLTGSSERECQGNGVWSGTEPICRQPYSYDFPEDVAPALGTSFSHMLGATNPTQKTKESLGRKIQIQRSGHLNLYLLLDCSQSVSENDFLIFKESASLMVDRIFSFEINVSVAIITFASEPKVLMSVLNDNSRDMTEVISSLENANYKDHENGTGTNTYAALNSVYLMMNNQMRLLGMETMAWQEIRHAIILLTDGKSNMGGSPKTAVDHIREILNINQKRNDYLDIYAIGVGKLDVDWRELNELGSKKDGERHAFILQDTKALHQVFEHMLDVSKLTDTICGVGNMSANASDQERTPWHVTIKPKSQETCRGALISDQWVLTAAHCFRDGNDHSLWRVNVGDPKSQWGKEFLIEKAVISPGFDVFAKKNQGILEFYGDDIALLKLAQKVKMSTHARPICLPCTMEANLALRRPQGSTCRDHENELLNKQSVPAHFVALNGSKLNINLKMGVEWTSCAEVVSQEKTMFPNLTDVREVVTDQFLCSGTQEDESPCKGESGGAVFLERRFRFFQVGLVSWGLYNPCLGSADKNSRKRAPRSKVPPPRDFHINLFRMQPWLRQHLGDVLNFLPL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service