Skip to Content
Merck
All Photos(4)

Key Documents

PLA0044

Sigma-Aldrich

Rabbit anti-Sp1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

specificity protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

mouse, human

technique(s)

ChIP: 4-30 μg
immunohistochemistry: 1:2,000-1:10,000
immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000-1:10,000

accession no.

XP_028606.4

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... Sp1(6667)

Immunogen

The epitope recognized by PLA0044 maps to a region between residue 750 and the C-terminus (residue 785) using the numbering given in entry XP_028606.4 (GeneID 6667).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Specificity protein 1 (Sp1) is the original member of the Sp1-like/KLF family of zinc-finger transcription factors that binds GC-rich DNA elements to regulate transcription of genes involved in a variety of cellular functions. Sp-1 interacts with coactivators such as p300/CBP, TAFII130, and CRSP.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Qiu et al.
Biochimica et biophysica acta, 1840(7), 2244-2252 (2014-03-07)
Cellular nucleic acid binding protein (CNBP) has been implicated in vertebrate craniofacial development and in myotonic dystrophy type 2 (DM2) and sporadic inclusion body myositis (sIBM) human diseases by controlling cell proliferation and survival to mediate neural crest expansion. CNBP
Hui-Hui Cao et al.
PloS one, 9(8), e106007-e106007 (2014-08-26)
Esophageal squamous cell carcinoma (ESCC) has the highest mortality rates in China. The 5-year survival rate of ESCC remains dismal despite improvements in treatments such as surgical resection and adjuvant chemoradiation, and current clinical staging approaches are limited in their
Deepti Malik et al.
Molecular cancer, 13, 175-175 (2014-07-20)
microRNAs (miRNAs) play both oncogenic and oncostatic roles in leukemia. However, the molecular details underlying miRNA-mediated regulation of their target genes in pediatric B- and T-cell acute lymphoblastic leukemias (ALLs) remain unclear. The present study investigated the relationship between miR-2909
Carolina Vizcaíno et al.
PloS one, 9(8), e104687-e104687 (2014-08-12)
Ovarian cancer has a poor prognosis due to intrinsic or acquired resistance to some cytotoxic drugs, raising the interest in new DNA-binding agents such as mithramycin analogues as potential chemotherapeutic agents in gynecological cancer. Using a genome-wide approach, we have
Luigi Formisano et al.
Journal of neuroscience research, 93(1), 167-177 (2014-08-06)
Polychlorinated biphenyls (PCBs) cause a wide spectrum of toxic effects in the brain through undefined mechanisms. Exposure to the PCB mixture Aroclor-1254 (A1254) increases the repressor element-1 silencing transcription factor (REST) expression, leading to neuronal death. This study sought to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service