Skip to Content
Merck
All Photos(1)

Key Documents

P0244

Sigma-Aldrich

Anti-PKR (ET-20) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Interferon-induced, double-stranded RNA-activated protein kinase (ET-20)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 68 kDa

species reactivity

human

technique(s)

microarray: suitable
western blot: 1:2,000 using a whole cell extract of the human epitheloid carcinoma HeLa cell line induced with interferon-γ

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EIF2AK2(5610)

General description

PKR is a double-stranded RNA-dependent serine/threonine kinase that regulates antiproliferative and antiviral functions of interferons. PKR also functions as a translational inhibitor. In genotoxic environments, PKR mediates CDC2 ubiquitination and G2 arrest in cells . Anti-PKR is antibody is specific for human PKR (68 kDa).

Immunogen

C-terminus of human PKR (amino acids 531-550), conjugated to KLH

Application

Anti-PKR (ET-20) antibody I suitable for use in western blot (1:2,000 using a whole cell extract of the human epitheloid carcinoma HeLa cell line induced with interferon-γ) and microarray.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New Cdc2 Tyr 4 phosphorylation by dsRNA-activated protein kinase triggers Cdc2 polyubiquitination and G2 arrest under genotoxic stresses.
Yoon, C., H., et al.
EMBO Reports, 393-9, 2010-2010 (2010)
K Kawakubo et al.
Virology, 264(1), 106-114 (1999-11-02)
The double-stranded RNA-dependent protein kinase PKR is an interferon-inducible enzyme that possesses antiviral and antiproliferative activities. We examined expression of PKR transcripts in human placenta tissue and cultured human amnion U cells. Alternative exon 2 structures were identified and characterized
H D Robertson et al.
Biochimie, 78(11-12), 909-914 (1996-01-01)
A model is presented for the regulation of the double-stranded RNA (dsRNA)-activated mammalian protein kinase PKR, which is involved in protein synthesis inhibition and the antiviral response in cells. A series of previous findings abut PKROs behavior are reviewed, including
Z Xu et al.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research, 18(8), 609-616 (1998-09-03)
The double-stranded RNA-dependent protein kinase (PKR) is a serine/threonine kinase that plays an important role in the antiviral activities of interferon (IFN). To determine the organization and regulation of the PKR locus, lambda phage and bacterial artificial chromosome (BAC) clones

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service