MIRRM02
MystiCq® microRNA® SYBR® Green qPCR ReadyMix™
with ROX™, formulation for miRNA RT-qPCR
Synonym(s):
SYBR® Green-based miRNA qPCR master mix, miRNA qPCR, microRNA qPCR master mix with ROX
About This Item
technique(s)
qPCR: suitable
compatibility
ABI 5700
ABI 7000
ABI 7300
ABI 7700
ABI 7900 HT
ABI 7900 HT Fast
ABI 7900
ABI StepOne
ABI StepOnePlus
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- Exclusively for use with the MystiCq microRNA qPCR Assay Primers, Control Primers, and Universal PCR primer
- 2X concentrated for easy reaction setup with hot-start Taq, dNTPs and optimized buffer
- Passive reference dye (if required) included at the optimal concentration
- This master mix can readily amplify fragments between 400 and 500 bp; however, for best results, amplicon size should be limited to 80 - 200 bp
Packaging
100RXN is packaged as 2 X 1.25 mL
500RXN is packaged as 10 X 1.25 mL
2000RXN is packaged as 1 X 50 mL
Other Notes
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Method for purification, reverse transcription and quantitative PCR for MicroRNAs using Mysticq reagents
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service