Key Documents
EP4925000022
Eppendorf® Reference® 2 G
single-channel, fixed, volume (2 μL), dark gray
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
single-channel, fixed
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 4925000022
volume
(2 μL)
color
dark gray
General description
Extraordinary precision and accuracy – Eppendorf´s premium pipette
Besides improved ergonomics the design focus lays on providing the most accurate results possible, optimum user safety, reliability and robustness for a long service life. The innovative single button operation is not only fast and easy, but it also allows active aerosol1) reduction and thus protects the user, the sample and the pipette. The sturdy handle manufactured with stainless steel protects the pipette from impact and in combination with the spring loaded bumpers the Reference 2 is likely to stay within calibration even if it takes a fall. These high levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducibility, Eppendorf′s Reference 2 is an ideal instrument for these liquid handling applications.
Single-Button Operation
  • Lower Operating forces for aspiration/dispensing/tip ejection limit strain on the user.
  • Allows users to move more quickly through their pipetting tasks by not having to move the thumb from side to side to eject tips.
  • With the ability to eject a tip immediately following dispensing, aerosol contamination of your pipette is considerably reduced.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)