Skip to Content
Merck
All Photos(1)

Key Documents

S7132

Supelco

Stanozolol

analytical standard

Synonym(s):

17β-Hydroxy-17α-methyl-5α-androstano[3,2-c]pyrazole, 5α-Androstane-17α-methyl-17β-ol-[3,2-c]pyrazole, Androstanazole, Stanazol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H32N2O
CAS Number:
Molecular Weight:
328.49
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

form

powder

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white to light yellow

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@]3([H])[C@]([H])(CC[C@@]4(C)[C@@]3([H])CC[C@]4(C)O)[C@@]1(C)Cc5cn[nH]c5C2

InChI

1S/C21H32N2O/c1-19-11-13-12-22-23-18(13)10-14(19)4-5-15-16(19)6-8-20(2)17(15)7-9-21(20,3)24/h12,14-17,24H,4-11H2,1-3H3,(H,22,23)/t14-,15+,16-,17-,19-,20-,21-/m0/s1

InChI key

LKAJKIOFIWVMDJ-IYRCEVNGSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

E Tudela et al.
Journal of chromatography. A, 1292, 195-200 (2013-02-05)
Stanozolol is one of the most frequently detected anabolic steroids in doping control samples. This compound is metabolized to a large extent and its metabolites can be detected in urine much longer than the parent compound. The main stanozolol metabolites
Unintended benefit of anabolic steroid use in hemophilia B leiden.
Emily K Rimmer et al.
American journal of hematology, 87(1), 122-123 (2011-11-01)
Laxmi S Inamdar Doddamani et al.
The Journal of steroid biochemistry and molecular biology, 129(3-5), 172-178 (2011-12-03)
The abuse of anabolic-androgenic steroids (AAS) for improved physical performance is associated with many deleterious effects. The present study aims to evaluate the short-term effect of an AAS compound stanozolol, on lipoprotein profile, granulopoiesis and immune response in adult female
S Ahi et al.
Drug testing and analysis, 1(11-12), 538-544 (2010-04-01)
The abuse of stanozolol is quite widespread in Indian sport. Its analysis is challenging and this has led to the development of new methods to improve its detection. A method was developed and validated for the detection of the three
Sule Unal et al.
Pediatric blood & cancer, 55(1), 174-176 (2010-03-09)
Severe essential cryofibrinogenemia is rare in childhood, and both the diagnosis and the management are challenging for pediatricians. An 11-year-old male, who had already lost two digits following cold exposure, was referred after multiple visits to various hospitals and subsequently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service