Skip to Content
Merck
All Photos(1)

Key Documents

15-1150

Sigma-Aldrich

Iron(III) chloride hexahydrate

SAJ first grade, ≥98.0%

Synonym(s):

Ferric chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3 · 6H2O
CAS Number:
Molecular Weight:
270.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥98.0%
grade:
SAJ first grade
form:
solid

grade

SAJ first grade

vapor pressure

1 mmHg ( 194 °C)

Assay

≥98.0%

form

solid

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

bp

280-285 °C (lit.)

mp

37 °C (lit.)

SMILES string

O.O.O.O.O.O.Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

NQXWGWZJXJUMQB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjay Kashyap et al.
ACS nano, 8(9), 9097-9106 (2014-08-28)
Biomineralization proteins are widely used as templating agents in biomimetic synthesis of a variety of organic-inorganic nanostructures. However, the role of the protein in controlling the nucleation and growth of biomimetic particles is not well understood, because the mechanism of
William H DePas et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(7), 2629-2634 (2013-01-30)
Bacterial biofilm formation is a complex developmental process involving cellular differentiation and the formation of intricate 3D structures. Here we demonstrate that exposure to ferric chloride triggers rugose biofilm formation by the uropathogenic Escherichia coli strain UTI89 and by enteric
D C Florian Wieland et al.
Colloids and surfaces. B, Biointerfaces, 109, 74-81 (2013-04-27)
The early stages of the formation of inorganic aggregates, composed of iron compounds at the solution-air interface, were investigated in situ. The properties of the solution-air interface were changed by using different Langmuir layers. In order to get insight into
Dattatraya H Dethe et al.
Organic letters, 15(3), 429-431 (2013-01-16)
A novel approach was developed for the synthesis of highly substituted indene derivatives, using an FeCl(3) catalyzed Prins-type cyclization reaction which was further applied in the total synthesis of jungianol and epi-jungianol.
Ko-Lin Kuo et al.
PloS one, 7(12), e50295-e50295 (2012-12-12)
The association between intravenous (IV) iron administration and outcomes in hemodialysis (HD) patients is still debated. Therefore, this study was aimed to assess the relationship between the IV administration of ferric chloride hexahydrate (Atofen®) and cardiovascular (CV) outcome and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service