Skip to Content
Merck
All Photos(1)

Key Documents

01-4680

Sigma-Aldrich

Ammonium fluoride

SAJ special grade, ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4F
CAS Number:
Molecular Weight:
37.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ special grade

Assay

≥97.0%

form

solid

availability

available only in Japan

pH

~6 (20 °C, 50 g/L)

SMILES string

N.F

InChI

1S/FH.H3N/h1H;1H3

InChI key

LDDQLRUQCUTJBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shanshan Wu et al.
Journal of colloid and interface science, 346(1), 12-16 (2010-03-12)
Fluorine-doped tin dioxide (FTO) nanocrystals were prepared with a sol-gel process followed by a hydrothermal treatment using SnCl(4) and NH(4)F as SnO(2) and fluorine dopant, respectively. The nanostructure and composition were characterized by X-ray diffraction (XRD), transmission electron microscopy (TEM)
L Touahir et al.
Bioelectrochemistry (Amsterdam, Netherlands), 80(1), 17-25 (2010-05-18)
(111) silicon surfaces can be controlled down to atomic level and offer a remarkable starting point for elaborating nanostructures. Hydrogenated surfaces are obtained by oxide dissolution in hydrofluoric acid or ammonium fluoride solution. Organic species are grafted onto the hydrogenated
Ethan Alden-Danforth et al.
Organic letters, 10(21), 4951-4953 (2008-10-15)
The catalytic, enantioselective, [4 + 2] cycloaddition reaction of ortho-quinone methides with silyl ketene acetals is described. This mechanistically interesting reaction, initiated by a chiral cinchona alkaloid-derived ammonium fluoride "precatalyst" complex, affords a variety of alkyl- and aryl-substituted 3,4-dihydrocoumarin products
Chitin-based organic networks: an integral part of cell wall biosilica in the diatom Thalassiosira pseudonana.
Eike Brunner et al.
Angewandte Chemie (International ed. in English), 48(51), 9724-9727 (2009-11-20)
Yuntao Li et al.
Journal of hazardous materials, 172(2-3), 635-640 (2009-08-12)
A F-doped vanadia/titania catalyst has been developed by partly substituting the lattice oxygen of the catalyst with fluorine, using NH(4)F as a precursor. The aim of this novel design was to promote the activity of a catalyst with low vanadia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service