Skip to Content
Merck
All Photos(1)

Key Documents

MABN128

Sigma-Aldrich

Anti-PITX3 Antibody, clone 12G4.1

clone 12G4.1, from mouse

Synonym(s):

Pituitary homeobox 3, Homeobox protein PITX3, Paired-like homeodomain transcription factor 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

12G4.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PITX3(5309)

General description

Pituitary homeobox 3 (UniProt O75364; also known as Homeobox protein PITX3, Paired-like homeodomain transcription factor 3) is encoded by the PITX3 (also known as ASMD, CTPP4, CTRCT11, PTX3) gene (Gene ID 5309) in human. PITX3 is a bicoid-related homeodomain transcription factor critical for the development of the ocular lens, mesencephalic dopaminergic neurons and skeletal muscle. PITX3 exerts its developmental impact on dopaminergic neurons by directly regulating the expression of a cascade of downstream genes, including TH, VMAT2, DAT, BDNF, and GDNF. PITX3 and the related PITX2 are both key regulators of intracellular redox state and play an important role in preventing DNA damage during fetal myogenesis. Human PITX3 gene mutations are linked to cataracts and anterior segment abnormalities and PITX3 polymorphisms are associated with Parkinson′s disease. In aphakia (ak) mice, deletions in the Pitx3 promoter region cause abnormal lens development.

Immunogen

KLH-conjugated linear peptide corresponding to a sequence within the N-terminal region of human PITX3.

Application

Anti-PITX3 Antibody, clone 12G4.1 is an antibody against PITX3 for use in Western Blotting.

Quality

Evaluated by Western Blotting in human retinoblastoma Y79 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected PITX3 in 10 µg of human retinoblastoma Y79 cell lysate.

Target description

~32 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service