Merck
56648-U
Supelco
HyperShear Ultra HPLC High Pressure Static Mixer
Binary tee housing assembly, volume 50 μL
About This Item
UNSPSC-Code:
41115854
Materialien
(wetted material 316 stainless steel/PEEK)
Beschreibung
Port size, 10-32 CPI (standard HPLC fittngs for 1/16" tubing)
Pressure rated to 18,000 psi
Volumen
50 μL
Allgemeine Beschreibung
HyperShear Mixers incorporate a highly efficient cross-flow shearing mechanism which produces vortex shear mixing over a wide flow range. This mixing technology typically delivers between 25% to 200% better mixing efficiency compared to conventional packed bed or tortuous path mixers.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)