Merck
56033
Omnifit® Verbinder mit Standardbohrung
Two-way valve connectors
About This Item
UNSPSC-Code:
12000000
Verpackung
pkg of 2 units
Hersteller/Markenname
Omnifit 1101
Methode(n)
LPLC: suitable
Bohrung
1.5 mm
Kompatibilität
for use with 1/16 in. tubing
Allgemeine Beschreibung
Omnifit® Standard Bore Connector is very versatile in nature and is used for connecting various types of hardwall tubings. The standard bore connector has a manual valve operation on each end, thus the flow can be switched on or off as required. It is primarily used for fluid-line interconnection, sampling and mixing.
Anwendung
Omnifit® Standard Bore Connector was used to construct the flow-injection system for amperometric determination of tacrine.
Leistungsmerkmale und Vorteile
O-Ringe und PTFE-Konus enthalten.
Rechtliche Hinweise
Omnifit is a registered trademark of Diba Industries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Lot/Batch Number
Leider sind derzeit keine COAs für dieses Produkt online verfügbar.
Die Dokumentenbibliothek aufrufen
Flow-Injection Amperometric Determination of Tacrine based on Ion Transfer across a Water-Plasticized Polymeric Membrane Interface.
Joaquin, A. C.
Sensors, 7, 1185-1192 (2007)
Modern Instrumental Techniques
Keith, A
Soil and Environmental Analysis, 1-700 (2003)