Merck
23238
Supelco
Schraubkappe
black phenolic hole cap (with polyseal liner), thread for 20-400, pkg of 144 ea
About This Item
UNSPSC-Code:
41122001
Materialien
black phenolic hole cap (with polyseal liner)
Verpackung
pkg of 144 ea
Fitting
thread for 20-400
Kompatibilität
for use with 30 mL (32 × 80 mm) ~ For Bottle (Volume and Dimensions): 60 mL (38 × 95 mm)
Allgemeine Beschreibung
Screw caps are available as hole caps for use with sealing septa. Black phenolic hole cap (with polyseal liner, thread 20-400) can withstand a temperature of 149°C.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)