Z740398
The SpecTainer™ II
size 120 mL, sterile
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
Empfohlene Produkte
Materialien
polyethylene cap
polypropylene container
Sterilität
sterile
Verpackung
pack of 300 ea
Hersteller/Markenname
Simport C567-120CYS
Größe
120 mL
Allgemeine Beschreibung
For applications needing the utmost security where sample integrity is of high importance. A great feature of the Spectainer I is that it incorporates a unique tamper evident screw cap ensuring peace of mind during transport or storage situations where someone might have tampered with the specimen.
Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used is free from latex. Containers are 95kpa compliant. Available in three sizes: 60, 90 and 120 ml. The 60 ml size however is not graduated. The sterile container protects its sterility with a tamper evident seal.
Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used is free from latex. Containers are 95kpa compliant. Available in three sizes: 60, 90 and 120 ml. The 60 ml size however is not graduated. The sterile container protects its sterility with a tamper evident seal.
Rechtliche Hinweise
SpecTainer is a trademark of Simport
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)