Merck
Z719579
Sorenson low binding standard tips
maximum volume 200 μL, pack of 1000 ea (bag), non-sterile
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.01
Materialien
colorless
polypropylene
Sterilität
non-sterile
Leistungsmerkmale
barrier: no
Verpackung
pack of 1000 ea (bag)
pack of 10000 ea (10 Bags of 1000 tips)
Hersteller/Markenname
Sorenson 10470T
Maximales Volumen
200 μL
Volumen
200 μL
Allgemeine Beschreibung
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.
Rechtliche Hinweise
Sorenson is a trademark of Sorenson BioScience, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)