Merck
Z709050
ART® non-filtered tip
HLT 300, size 1-300 μL, Pure packed, sterile
Synonym(e):
pipet tips, pipette tips
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.01
Sterilität
sterile
Leistungsmerkmale
DNase free
RNase free
Pure packed
Verpackung
pack of 960 ea
Hersteller/Markenname
Molecular BioProducts 3571
Maximales Volumen
300 μL
Größe
1-300 μL
Volumen
1—300 μL
Allgemeine Beschreibung
Ideal for Finnpipette and molded with reference marks at 10, 50 and 100 μl.
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
Rechtliche Hinweise
ART is a registered trademark of Molecular BioProducts, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)