Z688150
HandEvac handheld aspirator
hand operator with single channel adapter for disposable tips
Synonym(e):
Handheld Aspirator
Alle Fotos(1)
About This Item
UNSPSC-Code:
41105100
NACRES:
NB.22
Empfohlene Produkte
Hersteller/Markenname
Cole Parmer 1305035
Allgemeine Beschreibung
Rapidly remove supernatant from a variety of cell culture or multi-well plates and dishes!
This ergonomic, hand-held aspirator can be used with pasteur pipettes, plastic aspiration pipettes or a variety of adapters for specific applications. A pressure-sensitive button on the top of the unit controls the specific amount of vacuum applied. This prevents the accidental aspiration of cells or supernatant associated with normal vacuum techniques.
Use with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems. Autoclavable unit is supplied with a single-channel adapter for disposable pipette tips.
This ergonomic, hand-held aspirator can be used with pasteur pipettes, plastic aspiration pipettes or a variety of adapters for specific applications. A pressure-sensitive button on the top of the unit controls the specific amount of vacuum applied. This prevents the accidental aspiration of cells or supernatant associated with normal vacuum techniques.
Use with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems. Autoclavable unit is supplied with a single-channel adapter for disposable pipette tips.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)