Merck
Z373400
LiposoFast Liposome Factory
Basic unit with stabilizer
Synonym(e):
Factory for Liposomes, LiposoFast Kit, Liposome Factory
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.22
Beschreibung
Includes membrane support capsule, two 0.5 mL gas-tight syringes, benchtop stabilizer, and 50 polycarbonate membranes, 100 nm pore size.
Sterilität
non-sterile
Allgemeine Beschreibung
Fast, efficient formation of uniform-sized, unilamellar liposomes. The principle is simple: a lipid emulsion is repeatedly extruded through a porous polycarbonate membrane, forced back and forth by specially modified gas-tight syringes attached to the membrane support capsule. With the syringes provided, unit has 0.5 mL capacity and virtually zero dead volume. Membranes from 100 to 400 nm pore size available.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z373400-1EA4061835575428