Merck
Z371076
Sigma-Aldrich
Dialysis tubing closures
yellow, size 110 mm
About This Item
UNSPSC-Code:
41123102
NACRES:
NB.22
Materialien
yellow
Größe
110 mm
Allgemeine Beschreibung
Clamping closures gently lock over dialysis tubing, providing a leakproof seal without knots. Can be used with cellulose ester, regenerated cellulose, and PVDF dialysis membranes. Made entirely of Nylon 66, closures have an operating temperature range of 0-90°C, and can be sterilized with ethylene oxide, γ-irradiation, ethanol, or formalin.
Different colors can be used to code samples. For best results, select a closure 4-10 mm wider than the flat width of the dialysis tubing.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)