Z333905
BRAND® accu-jet® pro pipette controller membrane filters
spare membrane filters 0.2 μm, sterile
Synonym(e):
bottle top filters, laboratory filters, vacuum system filters
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Empfohlene Produkte
Materialien
PTFE
Sterilität
sterile
Leistungsmerkmale
hydrophobic
Verpackung
pkg of 1 ea
Hersteller/Markenname
BRAND 26530
Porengröße
0.2 μm pore size
Verwandte Kategorien
Allgemeine Beschreibung
The accu-jet®S / pro / macro pipette controller is designed to safeguard your equipment with its robust PP housing and integrated interior membrane that effectively blocks liquid. This protective feature ensures the longevity of your pipette controller while maintaining a clean and safe working environment.
Sonstige Hinweise
BR26530 (India)
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
accu-jet is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)