TMO4115-0500
Nalgene® sterile, single-use Erlenmeyer flask with vented closure
PETG, bottle volume 500 mL, case of 12 ea
Synonym(e):
Nalgene Erlenmeyer flask, single use plastic flask
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.13
Empfohlene Produkte
Materialien
PETG
Sterilität
sterile
Leistungsmerkmale
vented
Verpackung
case of 12 ea
Hersteller/Markenname
Nalgene 4115-0500
Flaschenvolumen
500 mL
Verwandte Kategorien
Allgemeine Beschreibung
Save preparation time and avoid contamination risk by using the Nalgene® Single-Use PETG Erlenmeyer Flasks, which are available with a flat or baffled bottom. These flasks are ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available with a vented or non-vented closure.
Leistungsmerkmale und Vorteile
- Sterile and single us, reducing contamination risk
- BPA-free
- Collapses when autoclaved, reducing bio hazardous waste
- Graduated for quick volume assessment
- Increases user safety due to the break-resistant and robust design of the crystal-clear PETG material
- Facilitates visual inspection and monitoring of contents due to its crystal-clear composition
- Equipped with a vented cap to support gas exchange during processes like cell culture
Sonstige Hinweise
Browse all Erlenmeyer flasks
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)