Merck
TMO362002-0008
Nalgene® Boston round bottle without cap
natural high-density polyethylene, volume 250 mL, case of 250 ea
Synonym(e):
HDPE bottle, nalgene bottle
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
natural high-density polyethylene
Verpackung
case of 250 ea
Hersteller/Markenname
Nalgene 362002-0008
Kapazität
250 mL
Volumen
250 mL
Allgemeine Beschreibung
Conduct demanding packaging applications with Thermo Scientific Nalgene Boston Round HDPE Bottles without Closure, which are both durable and reliable products. These high density polyethylene bottles are rigid and translucent with excellent chemical resistance. Uniform walls make these bottles resistant to splitting and puncturing.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)