Merck
TMO2425-0503
Nalgene® narrow-mouth Right-to-Know wash bottle
low-density polyethylene, bottle volume 500 mL, methanol, case of 24 ea
Synonym(e):
Nalgene bottle, plastic bottle
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
low-density polyethylene
narrow-mouth
Verpackung
case of 24 ea
Hersteller/Markenname
Nalgene 2425-0503
Flaschenvolumen
500 mL
Allgemeine Beschreibung
Meet regulatory labeling requirements for chemical containers with Thermo Scientific Nalgene Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable symbols and colors. These low density polyethylene bottles are flexible for easy dispensing and translucent for viewing contents.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries