OH30456387
Ohaus® Navigator® Portable Balance
Model NV1201, weighing capacity 1200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Alle Fotos(1)
About This Item
UNSPSC-Code:
41111502
NACRES:
NB.71
Empfohlene Produkte
Materialien
metal weighing plate
Leistungsmerkmale
CE compliant
Verpackung
pack of 1 ea
Hersteller/Markenname
Ohaus® 30456387
Messbereich Wiegekapazität
1200 g, resolution: 0.1 g
Waagschale B × L
14.4 cm × 19 cm
Allgemeine Beschreibung
OHAUS raises the bar in price-performance ratio with the newly-designed Navigator series which is perfect for general weighing applications in laboratory, industrial and education settings. Ruggedly constructed and multifunctional, the Navigator′s a powerful balance that can handle a diverse range of weighing applications with features such as fast stabilization time, overload protection at four times its rated capacity, and easy operation.
Anwendung
Weighing, Parts Counting, Percent Weighing, Checkweighing
Leistungsmerkmale und Vorteile
Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings
Rechtliche Hinweise
Navigator is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
OH30456387-1EA | 4065266751048 |