OH30073982
Ohaus® Pen pH Meter
Model ST10S
Alle Fotos(1)
About This Item
UNSPSC-Code:
41115603
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Ohaus® 30073982
Messbereich Salzgehalt
0.0-10.0 ppt
Akkuleistung
4 × 1.5 V, 200 hr life
Allgemeine Beschreibung
With durable IP67 waterproof ABS housing, protective sensor cap, and an automatic shutdown feature that preserves battery life, OHAUS pen meters can endure consistent use in rough, wet environments.
Anwendung
pH, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), salinity, total dissolved solids (TDS), temperature measurement
Leistungsmerkmale und Vorteile
Easily replaceable electrodes, automatic shut off feature
Rechtliche Hinweise
Ohaus is a registered trademark of Ohaus Corp.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)