Merck
HS15989R
Premium slide box
25 place, foam lining, white plastic
Synonym(e):
Schiebekasten, slide box microscope, slide storage, slide storage box
About This Item
UNSPSC-Code:
41123302
NACRES:
NB.17
Materialien
nickel closure
white plastic
Leistungsmerkmale
Holds 25 x slides 76x26 mm
Hersteller/Markenname
Heathrow Scientific HS15989R
L × B × H
141 × 88 × 35 mm
Leistungsmerkmale und Vorteile
Slide boxes are perfect for storage or transportation of standard size slides (100 (50 back to back) x 75 mm and 1 x 3").
  • Molded, rigid boxes securely hold slides in grooved slots
  • Durable ABS plastic boxes are available with cork lining only
  • Both the lining and the slots are numbered to correspond to the slide inventory sheet
  • Feature thumb-latch lock mechanism
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)