EP0030118421
Eppendorf Tube® Conical with snap cap
tube capacity 25 mL, sterile, pyrogen free, RNase and DNase free, RNase free, pkg of 150 pieces, bag of 6 × 25 tubes
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121703
NACRES:
NB.11
Empfohlene Produkte
Sterilität
sterile
Leistungsmerkmale
RNase and DNase free
RNase free
pyrogen free
Verpackung
case of 150 ea
pkg of 150 pieces
bag of 6 × 25 tubes
Hersteller/Markenname
Eppendorf® 0030118421
Röhrchenkapazität
25 mL
Verwandte Kategorien
Allgemeine Beschreibung
- Reduced height gives easy sample access even with low-volume pipettes and tips?–?reduced risk of cross-contamination between pipette and tube
- Single-handed opening and closing through SnapTec cap version for fast liquid extraction and addition
- Centrifugation stability up to 17,000 x g1) for both tube variants - SnapTec cap and screw cap
- Tightness of the lid is ensured over a broad temperature range
- Less storage space needed due to ~20 % lower height compared to 50 mL conical tubes
- Matching accessories for centrifugation, mixing and heating, sample preparation and storage ensure straightforward integration into existing laboratory environment
- Premium USP class 6 raw material and no use of slip agents, plasticizers, and biocides during manufacturing ensure highest sample integrity and optimal sample and pellet visibility
- Screw cap with flattened and grooved sides supports a user-friendly ergonomic handling, enables a stable upright positioning, and reduces contamination risk
- Sterile, pyrogen-, DNase-, RNase-, and DNA-free. Each individual lot is checked and certified for quality and purity.
- The 25 mL conical tube with SnapTec cap is autoclavable at 121 °C in 20 min.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Tube is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)