Merck
CLS70372160
Pyrex® Milk Dilution bottle with screw cap
capacity 160 mL, narrow mouth, graduated
Synonym(e):
Pyrex® glass bottles
About This Item
UNSPSC-Code:
41121800
Materialien
borosilicate glass bottle
clear bottle
narrow-mouth bottle
square bottle
Leistungsmerkmale
autoclavable
cap
soda lime glass
Verpackung
pkg of 12 ea
case of 48 ea
Hersteller/Markenname
Corning 70372-160
Kapazität
160 mL
Durchm. × H
44 mm × 150 mm
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)