Merck
CLS6621
Corning® Step-R Repeating Pipettor Syringe Tips
volume (0.5 mL), non-sterile
Synonym(e):
pipet tips
About This Item
UNSPSC-Code:
41121600
NACRES:
NB.02
Sterilität
non-sterile
Verpackung
pack of 100 ea
Hersteller/Markenname
Corning 6621
Volumen
(0.5 mL)
Allgemeine Beschreibung
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.