Merck
CLS6120P150
Corning® plain funnel, reusable
polypropylene
Synonym(e):
conical funnel, squibb separatory funnels
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.43
Materialien
polypropylene
Beschreibung
with short stem
Verpackung
pack of 12 ea
Hersteller/Markenname
Corning 6120P-150
Durchmesser
150 mm
Schaft L × H × AD
25 mm × 45 mm × 1.5 mm
Allgemeine Beschreibung
Durable polypropylene funnels are translucent and allow safe and easy transfers of solids or liquids from one vessel to another.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)