Merck
CLS4876
Corning® Transtar® liquid handling system
disposable reservoir liner, open
Synonym(e):
lab equipment accessory
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.11
Sterilität
sterile
Hersteller/Markenname
Corning 4876
Allgemeine Beschreibung
The Transtar is a portable, autoclavable liquid handling system designed for use with 96 well plates, A sterile 96 tip disposable cartridge, which loads into the Transtar system, enables liquids to be aspirated, transferred and dispensed over a volume range of 25 - 200 μL in 5 μL increments, Transtar accuracy us rated at 5% at all volume levels.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
TRANSTAR-96 is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)