Merck
CLS431446
Corning® cap, Erlenmeyer access, LUER, 2 L
Synonym(e):
Corning cap accessory
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Hersteller/Markenname
Corning 431446
Allgemeine Beschreibung
  • Aseptic transfer caps are chemically resistant and fit Corning® Erlenmeyer 2L flasks.
  • Features a venting port that ends in a 50mm diameter vent filter with a 0.2 μm pore Polyvinylidene fluoride (PVDF) membrane and a barbed fitting.
  • The liquid transfer port has an internal end with a rigid polypropylene dip tube angled to reach the bottom of the flask.
  • Heat sealable flexible tubing ends in a male Luer lock (MLL) closed with a female Luer sealing cap.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
CLS431446-6EA4061831708875