Merck
CLS11440
Corning® closed system Erlenmeyer flask
pre-assembled, sterile, capacity 1 L
Synonym(e):
conical flasks
About This Item
UNSPSC-Code:
41122104
NACRES:
NB.43
Materialien
flat bottom flask
orange polypropylene cap
polycarbonate
polypropylene cap
Sterilität
sterile
Leistungsmerkmale
cap
Verpackung
case of 4
Hersteller/Markenname
Corning 11440
Kapazität
1 L
Sonstige Hinweise
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.