BMSH3565180HD
Benchmark SureTherm 180L CO2 Incubator
AC/DC input 120 V, with High Heat Function
Synonym(e):
carbon dioxide incubator
About This Item
Empfohlene Produkte
Materialien
stainless steel
Leistungsmerkmale
with High Heat Function
Verpackung
pack of 1 ea
Hersteller/Markenname
Benchmark Scientific H3565-180HD
Parameter
temp. range at +5-60 °C (ambient)
0-20% (CO2 range)
inside W × L × H
47.3 cm (18.7 in.) × 52.8 cm (20.8 in.) × 71 cm (28 in.)
Verwandte Kategorien
Allgemeine Beschreibung
These are the first and only incubators with the optional (patent pending) IncuView LCI (Live Cell Imaging) technology for viewing cells in “real time”. An internal microscope and large, external display allow cells to be viewed without removal from the incubator, eliminating the inherent temperature fluctuations and risk of contamination. Cells are easily viewed on the large, integrated display or viewed locally or remotely with a wifi enabled phone, tablet or computer. Cell viewing requires the IncuView Integrated Microscope Shelf.
Leistungsmerkmale und Vorteile
- 6 sided heating
- Live cell imaging option
- proprietary heat distribution system
- high heat decontamination
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BMSH3565180HD | 4065269198505 |