BMSD113450
Benchmark Stainless Steel Bulk Beads
bead diam. 5 mm, acid washed
Alle Fotos(1)
About This Item
UNSPSC-Code:
41100000
NACRES:
NA.82
Empfohlene Produkte
Materialien
stainless steel
Verpackung
pack of 1000 ea
Hersteller/Markenname
Benchmark Scientific D1134-50
Bead-Durchm.
5 mm
Allgemeine Beschreibung
Beads are alcohol washed and heat treated to remove contaminants and render them nuclease and protease free
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)