Merck
AXYAP8200
AxyPet® pipette
8 channel, autoclavable, size 20-200 μL, pkg of 1 ea
Synonym(e):
pipet
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Leistungsmerkmale
8-channel
electronic: no
Verpackung
pkg of 1 ea
Hersteller/Markenname
Axygen Inc AP-8-200
Corning
Größe
20-200 μL
Rechtliche Hinweise
AxyPet is a registered trademark of Axygen, Inc.