Z763284
Aldrich® Sure/Seal™ joint adapter
with PTFE-faced EPDM rubber septum, PTFE body, joint: 14/20
Alle Fotos(2)
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.43
Empfohlene Produkte
Materialien
PTFE body
Verbindung
joint: 14/20
Verwandte Kategorien
Allgemeine Beschreibung
This adapter enables repeated cannulation into any jointed flask or tube using the Sure/Seal system of PTFE-faced EPDM rubber septum mounted in an all-PTFE body adapter. Complete adapter includes PTFE body, one septum, one polypropylene overcap with hole and plastic joint clip.
- Precision machined PTFE body with 14/20 or 24/40 joint and cap threads
- Replaceable PTFE-faced EPDM rubber septum has excellent resistance to solvents and reseals when punctured numerous times. Use with non-coring point needles to extend septum life
- Polypropylene over cap with hole compresses septum into adapter body to maintain seal
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z763284-1EA | 4061832122878 |