Merck
Z758841
Bollé Cobra wrap-around safety spectacles
ANSI compliant
Synonym(e):
protective eyewear, protective glasses, safety eyeglasses, safety eyewear, safety glasses, safety spectacles
About This Item
UNSPSC-Code:
46181802
Leistungsmerkmale
ANSI compliant
Verpackung
pkg of 1 ea
Hersteller/Markenname
Bollé 40037
Allgemeine Beschreibung
A multi-purpose wrap around safety goggle with a panoramic visual field. Enveloping and without any visual interference, the Cobra offers 180° panoramic visual field with perfect optical quality. The ability to interchange the temples with an adjustable strap and foam option makes the Cobra an ultra versatile model.
Cobra Kits to facilitate the optional strap conversion is available separately as: Z758868 or Z758876.
  • Upper protection
  • Interchangeable comfort foam
  • Anti-scratch, anti-fog, anti-static coatings
  • Non-slip bridge
  • Adjustable strap
Ähnliches Produkt
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)