Z743273
Julabo® adaptors
barbed fittings, for 12 mm i.d. tubing, pack of 2
Synonym(e):
barbed adaptor
Alle Fotos(1)
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.77
Empfohlene Produkte
Verpackung
pack of 2
Hersteller/Markenname
Julabo 8970445
Allgemeine Beschreibung
For use with Julabo® circulators, 2 barbed fittings for tubing 12 mm ID. For use with M16x1 fittings. Requires a female M16x1 cap nut.
Rechtliche Hinweise
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)