Merck
Z742792
Kern EG-N and WE-N series, loop for underfloor weighing
for balances with weigh plates 180 mm x 160 mm
About This Item
UNSPSC-Code:
41111502
NACRES:
NB.71
Hersteller/Markenname
Kern EG-A08
Allgemeine Beschreibung
Loop for underfloor weighing, suitable for precision balance KERN EW-N/EG-N (only for models with weighing plate size W×D 118 mm).
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)