Z742595
Accuris™ SmartDoc™ narrow band pass filter
535nm filter; Used for imaging green stains
Synonym(e):
Accuris™ SmartDoc™
Alle Fotos(1)
About This Item
UNSPSC-Code:
41106000
NACRES:
NB.24
Empfohlene Produkte
Leistungsmerkmale
535nm filter; Used for imaging green stains
Verpackung
pkg of 1
Hersteller/Markenname
Benchmark Scientific E5001-535
Allgemeine Beschreibung
A narrow band pass filter of 535 nm used for imaging green stains on a UV transilluminator.
Rechtliche Hinweise
Accuris is a trademark of Benchmark Scientific, Inc.
SmartDoc is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z742595-1EA | 4061826162064 |