Merck
Z679089
Tamper evident caps
for 5, 10, and 12 mL tubes
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.11
Verpackung
pack of 1000 ea
Hersteller/Markenname
Simport T552-WTP
Allgemeine Beschreibung
Tamper and leak-roof polyethylene caps with specially designed lip seal, guarantee the utmost security and sample integrity, non-sterile.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)