Z615692
Julabo® bath test tube racks
to fit 90 microtiter tubes, for water baths, diam. 11-12 mm
Synonym(e):
tube holder
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.77
Empfohlene Produkte
Materialien
polypropylene
Leistungsmerkmale
Holds 90 x 11-13 mm tubes
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 8970382
Parameter
+80 °C temp. limit
Durchmesser
11-12 mm
Verwandte Kategorien
Allgemeine Beschreibung
Polypropylene tray and test tube inserts can be used up to +80 °C. When tempering solvent-containing or acidic samples above 60 °C, it is recommended using stainless steel test tube inserts, as the plastic can be attacked.
Rechtliche Hinweise
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z615692-1EA | 4061833062548 |