Z567574
Aldrich® Benchclamp™ 3-prong
dual adjustment grippy knobs, size S
Synonym(e):
bench clamps
Alle Fotos(2)
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.35
Empfohlene Produkte
Größe
S
Backenkapazität
0-30 mm
Zinke B
6 mm
Verwandte Kategorien
Allgemeine Beschreibung
Aldrich benchclamp clamps have a nickel-plated finish and are corrosion resistant. The clamps come with slip-on fiberglass finger covers. The Grippy Knob design has a round nut which is easy to turn making tightening the clamps around glassware fast and convenient.
Round extension arms allow the clamps to be rotated 360°. Extension arms allow placement of apparatus at various distances from lab frames. Supplied with vinyl and fiberglass prong covers for high temperature applications.
NOTE: An additional clamp holder is required to attach clamps to frames or other apparatus.
Round extension arms allow the clamps to be rotated 360°. Extension arms allow placement of apparatus at various distances from lab frames. Supplied with vinyl and fiberglass prong covers for high temperature applications.
NOTE: An additional clamp holder is required to attach clamps to frames or other apparatus.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchclamp is a trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z567574-1EA | 4061837637063 |