Z402362
Best® butyl gloves
size XL
Alle Fotos(1)
About This Item
UNSPSC-Code:
46181504
NACRES:
NB.35
Empfohlene Produkte
Materialien
black gloves
butyl rubber
Größe
XL
Sterilität
non-sterile
Leistungsmerkmale
latex free
powder-free
reusable
Verpackung
pkg of 1 pair
Hersteller/Markenname
Best Manufacturing 878-10
L × Dicke
350 mm × 0.70 mm
Leistungsmerkmale und Vorteile
- Impermeable
- Resistant to water
- Great tactility and performance
- Provides permeation resistance to gases and water vapors
- Unsupported, heavy-duty, 100% butyl rubber
- Injection molded
- Smooth grip with rolled cuff
- Offers flexibility and durability
- Excellent resistance to ketones, acids, caustics, isocyanate, and gases
Rechtliche Hinweise
Best is a registered trademark of Showa Glove Co.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z402362-1PAK | 4061834378242 |